Assay Details
Gene Symbol: SLC35A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001494564.5 | XP_001494614.1 | 2368 | 56 | ||
XM_005614132.3 | XP_005614189.1 | 2185 | 56 | |||
XM_014728841.2 | XP_014584327.1 | 1778 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
