Assay Details
Gene Symbol: FGF14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023621799.1 | XP_023477567.1 | 1430 | 75 | ||
XM_023621800.1 | XP_023477568.1 | 731 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
