Assay Details
Gene Symbol: PTCHD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023625059.1 | XP_023480827.1 | 603 | 73 | ||
XM_023625061.1 | XP_023480829.1 | 675 | 73 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
