Assay Details
Gene Symbol: PTCHD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023625059.1 | XP_023480827.1 | 1085 | 74 | ||
XM_023625061.1 | XP_023480829.1 | 1157 | 74 | |||
XM_023625062.1 | XP_023480830.1 | 911 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
