Assay Details
Gene Symbol: LIME1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023626955.1 | XP_023482723.1 | 699 | 101 | ||
XM_023626956.1 | XP_023482724.1 | 817 | 101 | |||
XM_023626957.1 | XP_023482725.1 | 617 | 101 |

More Information
Set Membership: |
![]() ![]() |
