Assay Details
Gene Symbol: CSDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023631727.1 | XP_023487495.1 | 843 | 62 | ||
XM_023631728.1 | XP_023487496.1 | 926 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
