Assay Details
Gene Symbol: NEIL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023636266.1 | XP_023492034.1 | 793 | 109 | ||
XM_023636267.1 | XP_023492035.1 | 879 | 109 | |||
XM_023636268.1 | XP_023492036.1 | 627 | 109 | |||
XM_023636269.1 | XP_023492037.1 | 605 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
