Assay Details
Gene Symbol: TGFBR3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023644657.1 | XP_023500425.1 | 1480 | 103 | ||
XM_023644658.1 | XP_023500426.1 | 543 | 103 |

More Information
Set Membership: |
![]() ![]() |
