Assay Details
Gene Symbol: TGFBR3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023644657.1 | XP_023500425.1 | 1919 | 78 | ||
XM_023644658.1 | XP_023500426.1 | 982 | 78 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: TGFBR3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023644657.1 | XP_023500425.1 | 1919 | 78 | ||
XM_023644658.1 | XP_023500426.1 | 982 | 78 |