Assay Details
Gene Symbol: SRRD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023646935.1 | XP_023502703.1 | 261 | 87 | ||
XM_023646936.1 | XP_023502704.1 | 261 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() |
