Assay Details
Gene Symbol: PYCR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023649121.1 | XP_023504889.1 | 209 | 78 | ||
XM_023649122.1 | XP_023504890.1 | 209 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
