Assay Details
Gene Symbol: EXOC3L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023649896.1 | XP_023505664.1 | 2178 | 56 | ||
XM_023649897.1 | XP_023505665.1 | 2002 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() |
