Assay Details
Gene Symbol: FLT3LG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023650136.1 | XP_023505904.1 | 318 | 70 | ||
XM_023650137.1 | XP_023505905.1 | 320 | 70 | |||
XM_023650138.1 | XP_023505906.1 | 323 | 70 | |||
XM_023650139.1 | XP_023505907.1 | 1014 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
