Assay Details
Gene Symbol: TK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023651911.1 | XP_023507679.1 | 695 | 59 | ||
XM_023651912.1 | XP_023507680.1 | 619 | 59 | |||
XM_023651913.1 | XP_023507681.1 | 537 | 59 | |||
XM_023651914.1 | XP_023507682.1 | 1918 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() |
