Assay Details
Gene Symbol: EIF5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023653443.1 | XP_023509211.1 | 374 | 65 | ||
XM_023653444.1 | XP_023509212.1 | 475 | 65 | |||
XM_023653445.1 | XP_023509213.1 | 301 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
