Assay Details
Gene Symbol: TPPP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001496430.5 | XP_001496480.2 | 479 | 74 | ||
XM_005608330.3 | XP_005608387.1 | 420 | 74 | |||
XM_023638668.1 | XP_023494436.1 | 404 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
