Assay Details
Gene Symbol: GLRA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001498316.4 | XP_001498366.3 | 1402 | 77 | ||
XM_014737750.2 | XP_014593236.1 | 1402 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
