Assay Details
Gene Symbol: LCAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001498463.5 | XP_001498513.1 | 227 | 57 | ||
XM_005608349.3 | XP_005608406.1 | 227 | 57 | |||
XM_005608350.3 | XP_005608407.1 | 227 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() |
