Assay Details
Gene Symbol: SLA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001499435.5 | XP_001499485.1 | 1101 | 60 | ||
XM_014735136.2 | XP_014590622.1 | 1259 | 60 | |||
XM_023626505.1 | XP_023482273.1 | 1209 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
