Assay Details
Gene Symbol: PPP1R1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001501286.3 | XP_001501336.1 | 366 | 81 | ||
XM_023622151.1 | XP_023477919.1 | 429 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() |
