Assay Details
Gene Symbol: TRAPPC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001503069.5 | XP_001503119.1 | 789 | 64 | ||
XM_023644956.1 | XP_023500724.1 | 789 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
