Assay Details
Gene Symbol: GABRA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001503397.5 | XP_001503447.3 | 482 | 91 | ||
XM_023617282.1 | XP_023473050.1 | 488 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() |
