Assay Details
Gene Symbol: PCP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005611498.3 | XP_005611555.1 | 1324 | 75 | ||
XM_014741374.2 | XP_014596860.2 | 880 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
