Assay Details
Gene Symbol: BSG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098795.2 | NP_001092265.1 | 3 - 4 | 326 | 62 | |
XM_023644130.1 | XP_023499898.1 | 400 | 62 | |||
GenBank mRNA | EF564280.1 | - | 3 - 4 | 327 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
