Assay Details
Gene Symbol: COMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081834.2 | NP_001075303.1 | 5 | 720 | 90 | |
NM_001114535.2 | NP_001108007.1 | 5 | 720 | 90 | ||
GenBank mRNA | AB178284.1 | - | 5 | 721 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
