Assay Details
Gene Symbol: AKR7A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_417628.6 | XP_417628.2 | 6 - 7 | 838 | 92 | |
GenBank mRNA | BX930804.2 | - | 6 - 7 | 711 | 92 | |
BX930844.1 | - | 6 - 7 | 828 | 92 | ||
BX931408.1 | - | 6 - 7 | 807 | 92 | ||
BX934026.2 | - | 6 - 7 | 830 | 92 | ||
BX934046.1 | - | 6 - 7 | 830 | 92 | ||
CR353499.1 | - | 6 - 7 | 825 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
