Assay Details
Gene Symbol: TPPP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_418894.6 | XP_418894.3 | 2 - 3 | 197 | 174 | |
GenBank mRNA | BX932523.2 | - | 2 - 3 | 162 | 174 | |
BX932684.1 | - | 2 - 3 | 182 | 174 | ||
BX934333.2 | - | 2 - 3 | 186 | 174 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_418894.6 transcripts.
Set Membership: |
![]() ![]() ![]() |
