Assay Details
Gene Symbol: RPS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277164.1 | NP_001264093.1 | 3 - 4 | 273 | 131 | |
GenBank mRNA | BX933609.1 | - | 2 - 3 | 259 | 131 | |
BX934117.1 | - | 2 - 3 | 256 | 131 | ||
BX934597.2 | - | 2 - 3 | 259 | 131 | ||
BX935640.2 | - | 2 - 3 | 259 | 131 | ||
CN226583.1 | - | 2 - 3 | 264 | 131 | ||
CR386179.1 | - | 2 - 3 | 163 | 131 | ||
CR386183.1 | - | 2 - 3 | 255 | 131 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
