Assay Details
Gene Symbol: VPS29L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277505.1 | NP_001264434.1 | 1 - 2 | 40 | 64 | |
GenBank mRNA | BX935605.1 | - | 1 - 2 | 41 | 64 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001277505.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
