Assay Details
Gene Symbol: MRAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001320907.1 | NP_001307836.1 | 1 - 2 | 133 | 69 | |
XM_015284715.2 | XP_015140201.1 | 5 - 6 | 671 | 69 | ||
XM_025148762.1 | XP_025004530.1 | 4 - 5 | 1237 | 69 | ||
XM_025148763.1 | XP_025004531.1 | 5 - 6 | 640 | 69 | ||
XM_025148764.1 | XP_025004532.1 | 5 - 6 | 601 | 69 | ||
XM_025148765.1 | XP_025004533.1 | 5 - 6 | 868 | 69 | ||
GenBank mRNA | BX929558.1 | - | 2 - 3 | 187 | 69 | |
BX936128.1 | - | 5 - 6 | 663 | 69 | ||
CR405797.1 | - | 3 - 4 | 353 | 69 | ||
KT183012.1 | - | 1 - 2 | 133 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
