Assay Details
Gene Symbol: MSRB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135558.2 | NP_001129030.1 | 1 - 2 | 92 | 83 | |
NM_001346746.2 | NP_001333675.2 | 1 - 2 | 92 | 83 | ||
GenBank mRNA | AJ454103.1 | - | 1 - 2 | 93 | 83 | |
BG710256.1 | - | 1 - 2 | 72 | 83 | ||
BM487648.1 | - | 1 - 2 | 72 | 83 | ||
BU220886.1 | - | 1 - 2 | 60 | 83 | ||
BX930278.2 | - | 1 - 2 | 35 | 83 | ||
BX950711.1 | - | 1 - 2 | 84 | 83 | ||
BX950828.2 | - | 1 - 2 | 60 | 83 | ||
CD761648.1 | - | 1 - 2 | 109 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
