Assay Details
Gene Symbol: HDHD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_414700.6 | XP_414700.4 | 2 - 3 | 142 | 67 | |
GenBank mRNA | BX929292.2 | - | 2 - 3 | 122 | 67 | |
BX950351.2 | - | 2 - 3 | 120 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
