Assay Details
Gene Symbol: LLGL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025141655.1 | XP_024997423.1 | 1 - 2 | 270 | 73 | |
GenBank mRNA | BX930166.2 | - | 1 - 2 | 275 | 73 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_025141655.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
