Assay Details
Gene Symbol: LLGL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015295434.2 | XP_015150920.1 | 2 - 3 | 151 | 71 | |
XM_015295435.2 | XP_015150921.1 | 2 - 3 | 151 | 71 | ||
XM_015295436.2 | XP_015150922.1 | 4 - 5 | 810 | 71 | ||
XM_015295438.2 | XP_015150924.1 | 3 - 4 | 763 | 71 | ||
XM_025141654.1 | XP_024997422.1 | 2 - 3 | 151 | 71 | ||
XM_025141655.1 | XP_024997423.1 | 2 - 3 | 375 | 71 | ||
XM_025141656.1 | XP_024997424.1 | 3 - 4 | 274 | 71 | ||
GenBank mRNA | BX930166.2 | - | 2 - 3 | 380 | 71 | |
BX931931.2 | - | 2 - 3 | 168 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
