Assay Details
Gene Symbol: RNF219 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015277350.2 | XP_015132836.1 | 6 | 2746 | 131 | |
XM_025146765.1 | XP_025002533.1 | 6 | 1859 | 131 | ||
XM_025146766.1 | XP_025002534.1 | 6 | 1840 | 131 | ||
XM_416999.6 | XP_416999.5 | 6 | 1869 | 131 | ||
GenBank mRNA | BX934734.2 | - | 1 | 1044 | 131 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
