Assay Details
Gene Symbol: PTCHD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015285145.2 | XP_015140631.1 | 1 - 2 | 1314 | 80 | |
GenBank mRNA | BX934281.2 | - | 1 - 2 | 588 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
