Assay Details
Gene Symbol: PLBD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015294865.2 | XP_015150351.1 | 11 - 12 | 1485 | 72 | |
XM_015294866.2 | XP_015150352.1 | 9 - 10 | 1299 | 72 | ||
GenBank mRNA | CR407176.1 | - | 2 - 3 | 234 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
