Assay Details
Gene Symbol: MTSS1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015279207.2 | XP_015134693.1 | 7 - 8 | 707 | 74 | |
XM_015279208.2 | XP_015134694.1 | 7 - 8 | 707 | 74 | ||
XM_015279212.2 | XP_015134698.1 | 7 - 8 | 707 | 74 | ||
GenBank mRNA | CR407184.1 | - | 7 - 8 | 441 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
