Assay Details
Gene Symbol: MTSS1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015279207.2 | XP_015134693.1 | 9 - 10 | 970 | 71 | |
XM_015279208.2 | XP_015134694.1 | 9 - 10 | 970 | 71 | ||
XM_015279210.2 | XP_015134696.1 | 8 - 9 | 961 | 71 | ||
XM_015279211.2 | XP_015134697.1 | 8 - 9 | 955 | 71 | ||
XM_015279212.2 | XP_015134698.1 | 9 - 10 | 970 | 71 | ||
XM_015279213.2 | XP_015134699.1 | 8 - 9 | 961 | 71 | ||
XM_025154251.1 | XP_025010019.1 | 9 - 10 | 964 | 71 | ||
GenBank mRNA | CR407184.1 | - | 9 - 10 | 704 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
