Assay Details
Gene Symbol: FOXRED2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001234651.5 | XP_001234652.1 | 4 - 5 | 1170 | 66 | |
XM_004937777.3 | XP_004937834.1 | 4 - 5 | 1189 | 66 | ||
GenBank mRNA | CR391328.1 | - | 1 - 2 | 213 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
