Assay Details
Gene Symbol: PUSL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277883.1 | NP_001264812.1 | 3 - 4 | 331 | 91 | |
NM_001277884.1 | NP_001264813.1 | 3 - 4 | 382 | 91 | ||
XM_015296879.2 | XP_015152365.1 | 3 - 4 | 344 | 91 | ||
XM_025142514.1 | XP_024998282.1 | 3 - 4 | 316 | 91 | ||
XM_025142515.1 | XP_024998283.1 | 3 - 4 | 330 | 91 | ||
GenBank mRNA | BX929622.1 | - | 3 - 4 | 382 | 91 | |
BX931895.1 | - | 3 - 4 | 331 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() |
