Assay Details
Gene Symbol: ECHDC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198652.3 | NP_001185581.1 | 2 - 3 | 301 | 71 | |
GenBank mRNA | BX931998.1 | - | 2 - 3 | 256 | 71 | |
BX932081.1 | - | 2 - 3 | 302 | 71 | ||
BX935838.2 | - | 2 - 3 | 245 | 71 | ||
CR353437.1 | - | 1 - 2 | 262 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
