Assay Details
Gene Symbol: EPFIP1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025149944.1 | XP_025005712.1 | 1 - 2 | 401 | 101 | |
GenBank mRNA | BX932745.1 | - | 1 - 2 | 96 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() |
