Assay Details
Gene Symbol: RIDA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001234688.5 | XP_001234689.4 | 1 - 2 | 97 | 97 | |
GenBank mRNA | BX932754.1 | - | 1 - 2 | 89 | 97 | |
BX935048.1 | - | 1 - 2 | 71 | 97 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001234688.5 transcripts.
Set Membership: |
![]() ![]() ![]() |
