Assay Details
Gene Symbol: BNIP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_421829.6 | XP_421829.3 | 2 - 3 | 272 | 70 | |
GenBank mRNA | BX931008.2 | - | 2 - 3 | 266 | 70 | |
BX932000.2 | - | 2 - 3 | 233 | 70 | ||
BX933976.1 | - | 2 - 3 | 211 | 70 | ||
BX933995.1 | - | 2 - 3 | 364 | 70 | ||
BX935850.2 | - | 2 - 3 | 241 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
