Assay Details
Gene Symbol: GUCY1A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025149959.1 | XP_025005727.1 | 2 - 3 | 490 | 102 | |
XM_420375.6 | XP_420375.1 | 3 - 4 | 605 | 102 | ||
GenBank mRNA | BX933178.2 | - | 3 - 4 | 538 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() |
