Assay Details
Gene Symbol: CWC15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277823.1 | NP_001264752.1 | 2 - 3 | 296 | 81 | |
NM_001277824.1 | NP_001264753.1 | 2 - 3 | 216 | 81 | ||
GenBank mRNA | AM067556.1 | - | 2 - 3 | 216 | 81 | |
BX932596.2 | - | 2 - 3 | 296 | 81 | ||
BX934467.2 | - | 2 - 3 | 189 | 81 | ||
BX936078.1 | - | 2 - 3 | 208 | 81 | ||
BX936226.2 | - | 2 - 3 | 217 | 81 | ||
CN219349.1 | - | 2 - 3 | 204 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() |
