Assay Details
Gene Symbol: EIF3I | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164395.1 | NP_001157867.1 | 9 - 10 | 858 | 69 | |
GenBank mRNA | CR352485.1 | - | 8 - 9 | 935 | 69 | |
CR385826.1 | - | 4 - 5 | 366 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
