Assay Details
Gene Symbol: PAPOLB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198601.3 | NP_001185530.1 | 15 - 16 | 1672 | 143 | |
XM_015287288.2 | XP_015142774.1 | 15 - 16 | 1679 | 143 | ||
XM_015287291.2 | XP_015142777.1 | 15 - 16 | 1679 | 143 | ||
GenBank mRNA | AF061758.1 | - | 11 - 12 | 1045 | 143 |

More Information
Set Membership: |
![]() ![]() ![]() |
