Assay Details
Gene Symbol: PAPOLB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198601.3 | NP_001185530.1 | 17 - 18 | 1939 | 99 | |
XM_015287288.2 | XP_015142774.1 | 17 - 18 | 1946 | 99 | ||
GenBank mRNA | AF061758.1 | - | 13 - 14 | 1312 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() |
